Learn More
Medchemexpress LLC Eltanexor Z-isomer | 1642300-78-4 | 96.0% | 428.29 | C17H10F6N6O | 10 MG

Supplier: Medchemexpress LLC HY100423A10MG
Eltanexor Z-isomer is the Z-isomer of the exportin-1 (XPO1) inhibitor Eltanexor, provided as a research-grade compound for preclinical pharmacology and biochemical studies of nuclear export inhibition and anticancer activity.
- High reported purity (95.98%).
- Chemical formula C17H10F6N6O; molecular weight 428.29.
- Available in 10 MG quantity for laboratory use.
- Recommended storage: powder and solvent stability under specified low-temperature conditions.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.