Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eltanexor Z-isomer | 1642300-78-4 | 96.0% | 428.29 g·mol⁻¹ | C17H10F6N6O | 25 MG
SDP

Supplier:  Medchemexpress LLC HY100423A25MG

Encompass_Preferred

Eltanexor Z-isomer is the less-active geometric isomer of the exportin-1 (XPO1/CRM1) inhibitor eltanexor, supplied for research use. The compound has documented in vitro activity across multiple cell lines and is offered as both solid material and ready-to-use DMSO solutions. Key analytical identifiers include a purity near 96%, molecular weight 428.29 g·mol⁻¹, and formula C17H10F6N6O.

  • Provided for research use only.
  • Reported purity of 95.98%.
  • Molecular weight 428.29 g·mol⁻¹ and formula C17H10F6N6O.
  • Available as solid and DMSO solution formats.
  • Suitable for in vitro assay development and pharmacology studies.
  • Documented activity across multiple cell lines.

Catalog No. 50-002-13895


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.