Learn More
Medchemexpress LLC Eltanexor Z-isomer | 1642300-78-4 | 96.0% | 428.29 g/mol | C17H10F6N6O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100423A5MG
Encompass_Preferred
Eltanexor Z-isomer is the Z-isomer of eltanexor, a second-generation exportin-1 (XPO1/CRM1) inhibitor supplied for research use only. It is provided as a solid and in DMSO solution formats and is intended for biochemical and preclinical studies of nuclear export inhibition and related cellular pathways.
  • Research-use only inhibitor isomer.
  • Available as solid and DMSO solution formats.
  • Commonly used in preclinical and biochemical studies of nuclear export inhibition.
  • Supplied with a CAS registry number for unambiguous identification.
  • Typical purity around 96%.
Catalog No. 50-002-14566