Learn More
Medchemexpress LLC Eltanexor Z-isomer | 1642300-78-4 | 96.0% | 428.29 g/mol | C17H10F6N6O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100423A50MG
Encompass_Preferred
Eltanexor Z-isomer is the less active geometric isomer of a CRM1 (exportin-1) inhibitor supplied for preclinical research. It is provided as a light yellow solid with documented purity and characterized molecular properties. The compound is used in vitro and in vivo to study nuclear export inhibition, comparative isomer activity, and combination therapy effects; storage and handling guidance is provided for both powder and solution formats.
  • Suitable for comparative and mechanism-of-action studies.
  • Provided as a solid with documented purity of 95.98%.
  • Available in multiple small research pack sizes and a solution format.
  • Stable under recommended storage conditions for powder and solvent preparations.
  • Includes CAS number, molecular formula, and molecular weight identifiers.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-14799