Learn More
Medchemexpress LLC ElteN378 | 1421366-99-5 | 100.0% | 378.46 g/mol | C23H26N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1457125MG
Encompass_Preferred
ElteN378 is a potent small-molecule inhibitor of FKBP12 used in preclinical research to probe FKBP-related pathways. It is applied in studies of neurodegenerative disorders, cell proliferation diseases, and cancer models. Supplied as a high-purity solid for laboratory use; handle according to institutional safety procedures.
  • Selective FKBP12 inhibition for target validation and mechanistic studies.
  • Suitable for in vitro and ex vivo assays.
  • High reported purity for reproducible results.
  • Soluble in common organic solvent DMSO for stock preparation.
  • Provided with CAS and analytical data for traceability.
Catalog No. 50-002-21567