Learn More
Medchemexpress LLC Eltoprazine hydrochloride | 98206-09-8 | 99.25% | 256.73 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16687A10MM/1ML
Encompass_Preferred
Eltoprazine hydrochloride is a 5-HT1A/5-HT1B receptors agonist and a 5-HT2C receptor antagonist. It shows antiaggressive and anxiogenic effects.
  • Acts as a 5-HT1A/5-HT1B receptors agonist and a 5-HT2C receptor antagonist.
  • Shows antiaggressive and anxiogenic effects.
  • For research use only.
Catalog No. 50-003-94627