Learn More
Medchemexpress LLC Eltoprazine hydrochloride | 98206-09-8 | 99.3% | 256.73 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16687A
Encompass_Preferred
Eltoprazine (DU 28853) hydrochloride is a 5-HT1A/5-HT1B receptors agonist and a 5-HT2C receptor antagonist. It shows antiaggressive and anxiogenic effects.
  • Acts as a 5-HT1A/5-HT1B receptors agonist and a 5-HT2C receptor antagonist.
  • Shows antiaggressive and anxiogenic effects.
  • Purity of 99.25%.
Catalog No. 50-003-95232