Learn More
Selleck Chemical LLC Eltrombopag 10mg 496775-61-2
SDP
Supplier:  Selleck Chemical LLC S450210MG
Encompass_Preferred
Eltrombopag, a member of the biarylhydrazone class, is a nonpeptide agonist of the thrombopoietin receptor (TpoR), used to treat chronic hepatitis C-associated thrombocytopenia and chronic immune (idiopathic) thrombocytopenia (ITP). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3890