Learn More
Medchemexpress LLC Eltrombopag | 496775-61-2 | MFCD20926253 | C25H22N4O4 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY15306200MG
Encompass_Preferred
Eltrombopag is an orally active thrombopoietin receptor nonpeptide agonist. It exhibits thrombopoietic activity and has been studied for low blood platelet counts in chronic immune thrombocytopenia. It is also used in cardiovascular research, shows strong inhibitory effects against multidrug-resistant Staphylococcus aureus, and can induce apoptosis in hepatocellular carcinoma (HCC).
  • Exhibits thrombopoietic activity
  • Used in research for low blood platelet counts in chronic immune thrombocytopenia
  • Utilized in cardiovascular research
  • Shows strong inhibitory effects against multidrug-resistant Staphylococcus aureus
  • Can induce apoptosis in hepatocellular carcinoma (HCC)
Catalog No. 50-003-72136