Learn More
Apexbio Technology LLC Eltrombopag Olamine 496775-62-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A33875.1
Encompass_Preferred
Eltrombopag Olamine is an orally bioavailable thrombopoietin receptor (c-mpl) agonist that directly stimulates platelet production through activation of the JAK2/STAT signaling pathway In cellular assays using murine BAF3 cells expressing human thrombopoietin receptor (hTpoR) eltrombopag induced STAT-dependent signaling responses with an EC50 value of approximately 0 27 M This compound promotes differentiation of human CD34 progenitor cells into megakaryocytes capable of platelet production Eltrombopag is commonly applied in hematological research particularly in studies involving thrombocytopenia and thrombopoiesis-related mechanisms
Catalog No. 50-000-62856