Learn More
Medchemexpress LLC Eltrombopag Olamine | 496775-62-3 | C29H36N6O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15306A50MG
Encompass_Preferred
Eltrombopag Olamine is an orally active, nonpeptide agonist of the thrombopoietin receptor, possessing thrombopoietic activity. It has been researched for treating low blood platelet counts associated with chronic immune thrombocytopenia and for cardiovascular research. It also exhibits strong inhibitory effects against multidrug-resistant *Staphylococcus aureus* and can induce apoptosis in hepatocellular carcinoma cells.
  • Orally active thrombopoietin receptor nonpeptide agonist
  • Used in research for low blood platelet counts associated with chronic immune thrombocytopenia
  • Relevant for cardiovascular research
  • Exhibits strong inhibitory effects against multidrug-resistant *Staphylococcus aureus*
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
Catalog No. 50-003-73492