Learn More
Medchemexpress LLC Elubrixin tosylate | 960495-43-6 | 98.1% | 635.51 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY18263C10MM/1ML
Encompass_Preferred
Elubrixin tosylate (SB-656933 tosylate) is a potent, selective, competitive, reversible, and orally active CXCR2 antagonist and an IL-8 receptor antagonist. It inhibits neutrophil CD11b upregulation (IC50 of 260.7 nM) and shape change (IC50 of 310.5 nM), making it suitable for research into inflammatory diseases.
  • Potent and selective CXCR2 and IL-8 receptor antagonist
  • Inhibits neutrophil CD11b upregulation
  • Inhibits neutrophil shape change
  • Orally active and reversible
  • Potential for inflammatory diseases research
Catalog No. 50-003-87672