Learn More
Glass Expansion Replacement barrel for Eluo nebulizer cleaning tool
SDP
Supplier:  Glass Expansion 7030058
Encompass_Preferred
Eluo Barrel. A replacement barrel for the Eluo Nebulizer Cleaning Tool (NC9462559, 70-ELUO). Try using Fluka RBS-25 Concentrate 1L (NC9000117, FLUKA25) with your Eluo, it is a concentrated cleaning solution that is highly effective in removing clogs from nebulizers, and deposits/residues from spray chambers, torches, injectors, and cones.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1906608