Learn More
Perkin Elmer US LLC Eluo Nebulizer Cleaner for Newer Nebulizers
Supplier:  Perkin Elmer US LLC N0777712
Encompass_Preferred
Previously there was no simple way to effectively clean and unblock nebulizers. Now, blocked nebulizers can be safely and easily restored to optimum performance with a revolutionary new cleaning instrument - the Eluo. The Eluo is designed to efficiently deliver a cleanser through the nebulizer capillary to dislodge particle build-up and thoroughly clean the nebulizer. Cleaning your nebulizer is now a simple three step process: Fill your Eluo with Methanol. Insert your nebulizer, tip first, into the Eluo. Clean your nebulizer by pressing the Plunger down. Use the Eluo regularly to maintain nebulizer performance and prolong nebulizer life. This version of the Eluo is effective for OpalMist, PolyCon, DuraMist, SeaSpray, and MEINHARD nebulizers.
Catalog No. 50-210-4214