Learn More
Glass Expansion Eluo Nebulizer Cleaner for Glass concentric Nebulizers (1161990, G3266-80020, 33010019, 220-95356-02)
SDP
Supplier:  Glass Expansion 70ELUO
Encompass_Preferred
Eluo Nebulizer Cleaner for Glass Concentric Nebulizers. Compatible with all Glass Expansion glass and quartz concentric nebulizers such as SeaSpray, MicroMist, Conikal, and Slurry. Used to clear nebulizer clogs. If the sample capillary becomes blocked with particulates, use the Eluo Nebulizer Cleaning Tool to backflush the blockage. The Eluo is only recommended for Glass Expansion glass and quartz concentric nebulizers. For OpalMist and DuraMist nebulizers, use the Eluo OPD version (501907353).If nebulizer blockages are frequent, consider adding the re-useable Guardian in-line filter (NC0897716). The Guardian in-line filter provides a simple and effective way to eliminate the risk of a blockage, and the filter can be cleaned using a simple attachment for the Eluo (NC0801128). Fluka RBS-25 Concentrate 1L (NC9000117) is an excellent cleaning solution for nebulizers and other sample introduction components.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC9462559
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.