Learn More
Electron Microscopy Sciences Aurion EM Anionic Gold Tracer, EM-grade 15 nm 10 ML
SDP
Supplier:  Electron Microscopy Sciences 25469
Encompass_Preferred
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones, cationic tracers bind to polyanionic moieties (membranes, acidic proteins). “Neutral” BSA coated gold tracers are useful for detecting open connections and tissue damage. Under proper conditions and with suitable microscopical techniques the tracers can also be used to follow cellular events in time.
Particle size: EM-grade 15 nm
Catalog No. 50-245-49