Learn More
Selleck Chemical LLC Emapunil (AC 5216) S2592-5mg
SDP
Supplier:  Selleck Chemical LLC S25925MG
Encompass_Preferred
Emapunil (AC-5216 18 kiloDalton Translocator Protein XBD173) is a novel mitochondrial benzodiazepine receptor/translocator protein (MBR/TSPO) ligand Emapunil(AC-5216) showes high affinity for MBRs prepared from rat whole brain with Ki of 0 297 nM and IC50 of 3 04 nM and 2 73 nM at rat glioma cells and human glioma cells respectively
Catalog No. 50-311-1883