Learn More
Selleck Chemical LLC Emb-02-A4060-1MG
SDP
Supplier:  Selleck Chemical LLC A40601MG
Encompass_Preferred
Emb-02 is a bispecific antibody targeting the immune checkpoints PD-1 and LAG-3 with potential antitumor effects It restores effector T cell function and enhances antitumor activity M W 146 82 kDa
Catalog No. 50-313-6381