Learn More
Selleck Chemical LLC Emb-06-A4040-1MG
SDP
Supplier:  Selleck Chemical LLC A40401MG
Encompass_Preferred
Emb-06 is a tetravalent CD3 BCMA bispecific antibody in a Fabs-In-Tandem-Ig format with four active antigen-binding sites It targets BCMA on tumor cells and CD3 on T cells promoting immune activation and antitumor activity M W 241 36 kDa
Catalog No. 50-313-8493