Learn More
Medchemexpress LLC (+)-EMD 57033 | 147527-31-9 | 99.7% | 425.50 g/mol | C22H23N3O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY106844A10MG
Encompass_Preferred
(+)-EMD 57033 is a research-grade small-molecule cardiac troponin C (cTnC) activator and Ca2+ sensitizer supplied as the (+) enantiomer for in vitro and biochemical research. It is provided as a solid powder with documented analytical data and is not for human use.
  • Activates cardiac troponin C and increases calcium sensitivity.
  • Supplied as the (+) enantiomer for stereospecific studies.
  • High purity suitable for biochemical and pharmacological assays.
  • Provided as a solid powder with defined storage recommendations.
  • Molecular weight 425.50 g/mol; formula C22H23N3O4S.
  • Available in milligram quantities for laboratory research.
Catalog No. 50-002-41148