Learn More
Medchemexpress LLC EMD-95885 | 178165-43-0 | 382.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY170410100MG
Encompass_Preferred
EMD-95885 is a selective NR2B-containing NMDA receptors antagonist with an IC₅₀ of 3.9 nM. It does not interact with other sites on the NMDA receptor.
  • Target: iGluR
  • Pathway: Membrane transporter/ion channel; neuronal signaling
Catalog No. 50-003-83896