Learn More
Medchemexpress LLC Emd534085 | 858668-07-2 | 99.55% | 476.53 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15000100MG
Encompass_Preferred
EMD534085 is a potent and selective inhibitor of the mitotic kinesin-5 with an IC50 of 8 nM. It is intended for research use only. This product offers:
  • Potently and selectively inhibits mitotic kinesin-5.
  • Does not inhibit other tested kinesins at 1 μM or 10 μM.
  • Binds to the allosteric pocket of kinesin-5.
  • Induces rapid cell death in HL60 cells during mitotic arrest.
  • Exhibits antiproliferative activity against human HCT116 cells with an IC50 of 30 nM.
  • Leads to significant tumor growth reduction in mice without toxic side effects.
Catalog No. 50-003-60243