Learn More
Medchemexpress LLC EMD638683 | 1181770-72-8 | C18H18F2N2O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15193
Encompass_Preferred
EMD638683 (HY-15193) is an orally active SGK1 inhibitor with an IC50 of 3 μM. It strongly inhibits SGK1, moderately inhibits SGK2 and SGK3, and shows excellent selectivity for other AGC kinase family members. EMD638683 exhibits antihypertensive activity by inhibiting SGK1. It can also prevent heart inflammation and fibrosis caused by hypertension by inhibiting the cardiac NLRP3 inflammation body/IL-1β axis, independently of its blood pressure-lowering effect. This product promotes apoptosis of colon cancer cells and sensitizes radiotherapy.
  • Orally active SGK1 inhibitor
  • Strongly inhibits SGK1; moderately inhibits SGK2 and SGK3
  • High selectivity for other AGC kinase family members
  • Exhibits antihypertensive activity
  • Prevents heart inflammation and fibrosis caused by hypertension
  • Promotes apoptosis of colon cancer cells
  • Enhances sensitivity to radiotherapy
Catalog No. 50-003-75969