Learn More
Apexbio Technology LLC EMD638683 1181770-72-8 50mg
SDP
Supplier:  Apexbio Technology LLC A338950
Encompass_Preferred
EMD638683 (CAS 1181770-72-8) is a selective inhibitor of serum- and glucocorticoid-inducible kinases (SGKs) including isoforms SGK1 SGK2 and SGK3 SGK1 regulates key cellular processes such as ion transport survival proliferation and responses to steroid hormones EMD638683 shows inhibitory activity against SGK1 SGK2 and SGK3 (IC50 3 M) significantly reducing SGK-dependent phosphorylation of NDRG1 in vitro It demonstrates limited off-target interactions inhibiting only MSK1 and PRK2 at 1 M while sparing numerous other kinases In cell models EMD638683 influences mitochondrial polarization and caspase activity exhibiting antitumor effects in colorectal tumor cells and antihypertensive activity in vivo
Catalog No. 50-000-64275