Learn More
Cayman Chemical EmedastIn-13C-d3fumarate 5mg
SDP
Supplier:  Cayman Chemical 333785 MG
Encompass_Preferred
An internal standard for the quantification of emedastine by GC- or LC-MS
Catalog No. 50-295-0823