Learn More
Cayman Chemical EmeguIsIn A 100ug
SDP
Supplier:  Cayman Chemical 32929100 UG
Encompass_Preferred
A depsidone fungal metabolite; active against S. aureus, MRSA, and C. neoformans (MIC = 0.5 UG/ml for all) and P. falciparum (IC50 = 2.2 µM)
Catalog No. 50-294-8711