Learn More
Apexbio Technology LLC Emeramide 351994-94-0 100mg
SDP
Supplier:  Apexbio Technology LLC C6596100
Encompass_Preferred
Emeramide (CAS 351994-94-0) is a synthetic small molecule investigated for its chelating properties particularly in the context of heavy metal toxicity It functions as a metal-binding agent facilitating the sequestration and elimination of toxic metals such as mercury from biological systems Preclinical and clinical studies have examined its potential to mitigate mercury-induced toxicity by promoting metal excretion Emeramide has been utilized in research exploring therapeutic strategies for mercury poisoning supporting interest in its application as a chelator for heavy metal detoxification in biomedical research contexts
Catalog No. 50-000-88471