Learn More
Medchemexpress LLC Emi1 | 35773-42-3 | 99.8% | C20H18N2O3 | 10MG

Supplier: Medchemexpress LLC HY13807210MG
EMI1 is a small-molecule inhibitor selective for mutant epidermal growth factor receptor (EGFR) variants, including ex19del/T790M/C797S and L858R/T790M/C797S. It is intended for preclinical research into drug-resistant non-small-cell lung cancer (NSCLC) and is supplied with supporting product and safety documentation.
- Inhibits EGFR ex19del/T790M/C797S and L858R/T790M/C797S.
- Suitable for preclinical research on drug-resistant NSCLC.
- High chemical purity (≈99.8%).
- Offered as solid and as a 10 mM solution in DMSO.
- Molecular formula C20H18N2O3; molecular weight 334.37 g·mol-1.
- Product datasheet and SDS available for experimental use and safety guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.