Learn More
Medchemexpress LLC EMI48 | 34564-13-1 | 99.0% | 1 ML

Supplier: Medchemexpress LLC HY13106610MM/1ML
EMI48, a derivative of EMI1, exhibits enhanced potency against mutant EGFR compared to EMI1. It acts as an inhibitor of EGFR triple mutants, specifically EGFR ex19del/T790M/C797S and EGFR L858R/T790M/C797S. This compound is used in research for mutant EGFR-associated, drug-resistant non-small-cell lung cancer (NSCLC).
- Inhibits EGFR ex19del/T790M/C797S and EGFR L858R/T790M/C797S
- Enhanced potency toward mutant EGFR
- Purity: 99.02%
- Molecular weight: 348.40
- Formula: C21H20N2O3
- Appearance: Solid
- Color: Light yellow to yellow
- Soluble in DMSO (requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.