Learn More
ICHORBIO LTD Emicizumab Biosimilar - Research Grade
Supplier:  ICHORBIO LTD ICH51781MG
Encompass
Bulk emicizumab biosimilar. Available in 1mg, 5mg, 10mg, 20mg, 50mg and 100mg sizes. Low endotoxin, BSA and Azide Free. Research use only
Catalog No. NC3825214