Learn More
TARGETMOL CHEMICALS INC EMITEFUR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T272595MG
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Emitefur (BOF-A 2) is an orally active and potent 5-fluorouracil derivative with anticancer and antitumor activity.Emitefur is used in advanced gastric cancer breast cancer and metabolism-related diseases. purity: 98%
Catalog No. 50-258-0633