Learn More
Medchemexpress LLC Emixustat hydrochloride | 1141934-97-5 | MFCD28400873 | 99.3% | 299.84 | C16H26ClNO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY19720A10MG
Encompass_Preferred
Emixustat hydrochloride is a research-grade RPE65 inhibitor and visual cycle modulator used to study retinal physiology and degenerative eye diseases. It inhibits retinol isomerization with low-nanomolar potency (IC50 ~4.4 nM) and is supplied as a hydrochloride salt for stability; it is used in both in vitro and in vivo retinal research models.
  • Inhibits RPE65 with low-nanomolar potency.
  • Modulates the visual cycle to reduce retinol isomerization.
  • Suitable for in vitro and in vivo retinal research applications.
  • Supplied as a hydrochloride salt for improved handling and stability.
  • High purity by LCMS (reported >99%).
  • Available in small-mass packaging for dosing and preclinical studies.
Catalog No. 50-002-94627