Learn More
Medchemexpress LLC EML 425 | 1675821-32-5 | 98.0% | 440.49 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11026350MG
Encompass_Preferred
EML 425 is a potent and selective CREB binding protein (CBP)/p300 inhibitor with IC50s of 2.9 and 1.1 μM, respectively. It is a cell-permeable, selective, and noncompetitive inhibitor of KAT3 histone acetyltransferases, binding both the free enzyme and the enzyme-substrate complex. This compound induces a marked and time-dependent reduction in the acetylation of lysine H4K5 and H3K9 in U937 cells.
  • Potent and selective CREB binding protein (CBP)/p300 inhibitor
  • IC50s of 2.9 μM for CBP and 1.1 μM for p300
  • Cell-permeable
  • Selective
  • Noncompetitive versus both acetyl-CoA and a histone H3 peptide
  • Induces a marked and time-dependent reduction in the acetylation of lysine H4K5 and H3K9 in U937 cells
  • Alternative binding pocket near the substrate lysine binding groove and close to the acetylation site
  • Appearance: Solid
  • Color: Light yellow to yellow
Catalog No. 50-003-43703