Learn More
Medchemexpress LLC HY-110263 5mg Medchemexpress, EML 425 CAS:1675821-32-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1102635MG
Encompass_Preferred
Medchemexpress, HY-110263 5mg EML 425 CAS:1675821-32-5 EML425 is a potent and selective CREB binding protein (CBP)/p300 inhibitor with IC50s of 2.9 and 1.1 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8783