Learn More
TARGETMOL CHEMICALS INC Emlenoflast 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T92221MG
Encompass_Preferred
Emlenoflast (MCC7840) is a selective inhibitor of NLRP3 inflammasome. Purity 98.34%
Catalog No. 50-287-9327