Learn More
TARGETMOL CHEMICALS INC Emodepside 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T79015MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Emodepside (Bay 44-4400) is an anthelmintic drug that is effective against a number of gastrointestinal nematodes,with broad-spectrum anthelmintic activity. Purity 99.62%
Catalog No. 50-287-6333