Learn More
Medchemexpress LLC HY-14393 50mg Medchemexpress, Emodin CAS:518-82-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1439350MG
Encompass_Preferred
Medchemexpress, HY-14393 50mg Emodin CAS:518-82-1 Emodin (Frangula emodin), an anthraquinone derivative isolated from genus Rheum and Polygonum, is an anti-SARS-CoV compound. Emodin blocks the SARS coronavirus spike protein and angiotensin-converting enzyme 2 (ACE2) interaction. Emodin inhibits casein kinase-2 (CK2). Anti-inflammatory and anticancer effects. Emodin is a potent selective 11β-HSD1 inhibitor with the IC50 of 186 and 86 nM for human and mouse 11β-HSD1, respectively. Emodin ameliorates metabolic disorder in diet-induced obese mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1596