Learn More
Selleck Chemical LLC Empagliflozin (BI 10773) 5mg 864070-44-0
SDP
Supplier:  Selleck Chemical LLC S80225MG
Encompass_Preferred
Empagliflozin (BI-10773) is a potent and selective SGLT-2 inhibitor with IC50 of 3.1 nM, exhibits >300-fold selectivity over SGLT-1, 4, 5 and 6. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5565