Learn More
Cayman Chemical EmpaglIflozIn-d4 1mg
SDP
Supplier:  Cayman Chemical 223691 MG
Encompass_Preferred
An internal standard for the quantification of empagliflozin by GC- or LC-MS
Catalog No. 50-298-7941