Learn More
Medchemexpress LLC Empagliflozin (Standard) | 864070-44-0 | C23H27ClO7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15409R5MG
Encompass_Preferred
Empagliflozin (Standard) is an analytical standard of Empagliflozin, intended for research and analytical applications. It functions as a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor with an IC50 of 3.1 nM for human SGLT-2. This compound is used as a reference standard in assays.
  • Analytical standard for research and analytical applications
  • Selective sodium glucose cotransporter-2 (SGLT-2) inhibitor
  • Commonly utilized in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Features an IC50 of 3.1 nM for human SGLT-2
Catalog No. 50-003-76177