Learn More
Medchemexpress LLC Emprumapimod hydrochloride (PF-07265803 hydrochloride) | 1036404-17-7 | 98.0% | C24H30ClF2N5O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY145564A1MG
Encompass_Preferred
Emprumapimod (PF-07265803) hydrochloride is an orally active and selective inhibitor of p38α MAPK.
  • Can be used for the research of dilated cardiomyopathy.
  • Can be used for the research of acute inflammatory pain.
Catalog No. 50-003-55679