Learn More
Leica Biosystems EMPTY 30ML PJ - LABELED
Supplier:  Leica Biosystems 3800777
Encompass
Pre-filled Formalin Specimen Jars are impact-resistant, clear polypropylene jars prefilled with 10% Neutral, Phosphate Buffered Formalin. These wide-mouth jars provide easy access to specimens. The caps are ribbed, leakproof and easy to open or close even with wet hands or gloves. The amount of formalin in the specimen jars is approximately half the volume of the jar. Available empty, unlabeled or with current OSHA warning labels. Offered in a variety of sizes, including bulk packs.
Catalog No. NC2752761