Learn More
Selleck Chemical LLC Emricasan 25mg 254750-02-2 IDN-6556, PF 03491390, PF-03491390
SDP
Supplier:  Selleck Chemical LLC S777525MG
Encompass_Preferred
Emricasan (IDN-6556, PF 03491390, PF-03491390) is a potent irreversible pan-caspase inhibitor. Emricasan is an inhibitor of Zika virus infection. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5235