Learn More
Electron Microscopy Sciences EMS EMS-Hitachi SEM Mount Box
SDP
Supplier:  Electron Microscopy Sciences 76503
Encompass_Preferred
EMS introduces a unique box for Hitachi SEM mounts with M4 threaded hole in the base. The box is numbered 1 10 to accommodate Hitachi mounts from 15 to 32 mm diameter. The box can hold 10 x 15 mm mounts, 8 x 25 mm mounts or 2 x 32 mm mounts
Catalog No. 50-343-98