Learn More
Electron Microscopy Sciences EMS Tweezers Style 120, 150 mm
SDP
Supplier:  Electron Microscopy Sciences 78355120
Encompass_Preferred
Very well made tweezers that serves a wide range of applications, from electronics to dentistry and anatomy. Extremely accurate serration quality.
OAL: 150 mm
Tips: Strong and dull tips
Catalog No. 50-306-18