Learn More
Electron Microscopy Sciences EMS Tweezers Style 2AX, SA steel, Flat, round tips with self closing Tips
SDP
Supplier:  Electron Microscopy Sciences 783172AX
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Flat, round tips with self closing
OAL: 120 mm
Catalog No. 50-238-51