Learn More
Electron Microscopy Sciences EMS Tweezers Style 3C, SA steel, Very fine with superior finish Tips
SDP
Supplier:  Electron Microscopy Sciences 783183C
Encompass_Preferred
The tweezers are the Swiss highest quality. Recommended for handling of EM grids. Shorter than style 3. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very fine with superior finish
OAL: 110 mm
Catalog No. 50-238-48