Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Tweezers Style 5A, SA steel, Extra fine tips Tips
SDP

Supplier:  Electron Microscopy Sciences 783205A

Encompass_Preferred

The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Extra fine tips
OAL: 115 mm

Catalog No. 50-238-85


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.