Learn More
Electron Microscopy Sciences EMS Tweezers Style 7, Titanium Alloy, Fine Curved tips Tips
SDP
Supplier:  Electron Microscopy Sciences 783227TA
Encompass_Preferred
The tweezers are the Swiss highest quality. Titanium alloy has a relatively low density (4.5g/cm3), good mechanical properties and high use temperature (1600°F/870°C). It is non magnetic, has good corrosion resistance, and useful where a very high strength-to weight ratio is required.
Material: Titanium Alloy
Tips: Fine Curved tips
OAL: 115 mm
Catalog No. 50-238-94