Learn More
Electron Microscopy Sciences EMS UV/Cryo Chamber, 220 V
SDP
Supplier:  Electron Microscopy Sciences 7044520
Encompass_Preferred
The EMS UVC3 Cryo Chamber has been specifically designed to provide an economical yet precisely controlled low temperature environment for polymerization of polymers and resins for cryo specimen preparation. It can also be used for dehydration and infiltration of cryo specimens. This simple and functional unit has a large insulated chamber and comprises fully automatic temperature control for the polymerization process of embedding resins and polymers.
Temperature range can be set from -10 degree C to -40 degree C (14 degree F to -40 degree F
Catalog No. 50-930-302